Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 19/26 NS
Catalog # 201-3002
Supplier:   SciLabware
Multiple adapters, Quickfit®
Catalog # 201-3002
Supplier:  SciLabware
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 19/26 NS
Price
114,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With three necks. Two vertical and one angled at 45°.
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    19/26 NS