Fractionating columns, plain, Quickfit®
Supplier: SciLabware
Jump to:
About this item
PYREX®, borosilicate glass
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Ground glass joints for dependable performance. Designed for use with various types of column packing e.g. Raschig rings or Fenske helices.
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Ground glass joints for dependable performance. Designed for use with various types of column packing e.g. Raschig rings or Fenske helices.