Autoclave test packs, Bowie and Dick type, Comply™
Catalog # MMMEM1301
Fournisseur: 3M MEDICAL
Spécifications
- Type:Sterilization
- Colour:Blue
- Description:Bowie and Dick type autoclave test pack, Comply™, disposable, colour turns from light blue to pink (1301)
- Seq:02
- Fixture time:3,5 mins
- Operating temperature:< 230 °C
- Pk:20
Spécifications
À propos de cet article
With unique colour chemistry, the 3M™ Comply™ Disposable Bowie and Dick Type Autoclave Test Pack has been designed for single use in porous load sterilizers for daily Bowie and Dick tests.
- Easy to detect failures according to ISO 11140 Part 1
- Easy to interpret fail or pass
- Easy to locate and retrieve test sheet
- Easy to open "matchbox" system
- Paper indicator sheet
Bowie-Dick Test Packs are used to monitor the performance of vacuum-assisted pre-vac steam sterilizers; test packs will reproducibly detect residual air in the sterilizer chamber due to leaks, inadequate vacuum and non-condensable gases – any of which can compromise sterility.
Certifications: Conforms to BS EN 867-4 and carries the BSI Kite Mark.