Order Entry
Belgium
ContactUsLinkComponent
Disposable particle filtering respirators, P301, BizTex®
Disposable particle filtering respirators, P301, BizTex®
Catalog # PRTWP301WHR
Supplier:  Portwest
undefined
Disposable particle filtering respirators, P301, BizTex®
Catalog # PRTWP301WHR
Supplier:  Portwest

Specifications

  • Type:
    P301
  • Pk:
    10
  • With valve:
    +
  • Class:
    FFP3

Specifications

About this item

Disposable FFP3-mask consists of a PP substrate to which a laminated microporous polyethlene film is applied (PP/PE). The outer layer of fabric prevents the passage of liquid molecules inwards yet permits the vapour molecules that build up inside to pass through.

  • Adjustable nose bridge
  • Sweat absorbing nose foam (silicone/latex-free)
  • Twin elastic headband
  • Colour coded to indicate performance level
  • With easy flow exhalation valve
  • Can be used in concentrations of contaminats up to 20 times WEL
Certifications: EN 149:2001+A1:2009