ContactUsLinkComponent
Zoology: prepared slides with protozoa
Zoology: prepared slides with protozoa
Catalog # LIED74700
Supplier:  LIEDER JOHANNES
undefined
Zoology: prepared slides with protozoa
Catalog # LIED74700
Supplier:  LIEDER JOHANNES
Specifications
  • Description:
    Prepared slides with protozoa (10 pcs.)
  • Pk:
    1
Specifications
About this item
These prepared slides with protozoa include:
- Rhizopoda (Amoeba proteus) w.m.
- Radiolaria, mixed species, fossil
- Foraminifera from Mediterranean sea, mixed species, recent
- Euglena viridis, Flagellata, w.m.
- Ceratium hirundinella, fresh-water Dinoflagellate
- Trypanosoma gambiense, blood smear, causing African sleeping sickness
- Plasmodium, causes human malaria, blood smear
- Eimeria stiedae, infected liver, different phases of developing, sec.
- Paramecium, micro- and macronuclei stained, a common ciliate
- Vorticella, a coloniate ciliate