ContactUsLinkComponent
Sampling spoons, double-ended, SP Bel-Art
Sampling spoons, double-ended, SP Bel-Art
Catalog # BELAH367110000
undefined
Sampling spoons, double-ended, SP Bel-Art
Catalog # BELAH367110000
Specifications
  • Length:
    229 mm
  • Pk:
    1
  • Spoon size:
    13×25 mm
Specifications
About this item
Staninless steel, Fluo-Kem® PTFE coating. This spoon/spatula combination can be used for sampling and scraping.
  • Corrosion resistant stainless steel construction is strong and durable
  • Fluo-Kem® PTFE/FEP coating is FDA compliant and prevents material from sticking
  • Spatula size: 8×51 mm
Ideal for use in compact workstations or with balances.