Magnetic stirring bars, cylindrical, PTFE, Spinbar®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Stirring bars ensure centering and smooth running characteristics via a small removable pivot ring in the center. The pivot ring minimises the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers.
Frequently Bought Together