Order Entry
Belgium
ContactUsLinkComponent
Racks for standards bottles, SP Bel-Art
Racks for standards bottles, SP Bel-Art
Catalog # BELAF185132300
undefined
Racks for standards bottles, SP Bel-Art
Catalog # BELAF185132300

Specifications

  • Description:
    Rack for 30 USP standard bottles (23 mm)
  • Pk:
    1

Specifications

About this item

Acrylic, clear.

  • Compact footprint
  • Dimension: 330×110×220 mm