Order Entry
Belgium
ContactUsLinkComponent
Supports for round bottom flasks, SP Bel-Art
Supports for round bottom flasks, SP Bel-Art
Catalog # BELAF389500000
undefined
Supports for round bottom flasks, SP Bel-Art
Catalog # BELAF389500000

Specifications

  • Ext.Ø:
    171 mm
  • Description:
    Round-bottom flask support, Ø×H 171×51 mm
  • Height:
    51 mm
  • Pk:
    1

Specifications

About this item

This PP round bottom flask support fits any size round-bottom flask up to 10 liters using stepped, concentric rings.

  • Stackable
  • Hands-free, upright position
  • Good chemical and temperature resistance

Use several for adjustable work height.