ContactUsLinkComponent
Microcentrifuge tube floating racks, Prep-Safe™, SP Bel-Art
Microcentrifuge tube floating racks, Prep-Safe™, SP Bel-Art
Catalog # BELAF187420001
undefined
Microcentrifuge tube floating racks, Prep-Safe™, SP Bel-Art
Catalog # BELAF187420001
Specifications
  • Description:
    Prep-Safe™ microcentrifuge tube floating rack, natural, 28 place, PP
  • Pk:
    1
Specifications
About this item
PP
Unique tabs protect the samples from loss and contamination. Place tubes in slots and slide under tabs to keep tubes in place and caps closed when under pressure, floating or submerged. Use in water baths, ice or on the benchtop. Holds twenty eight 1,5 or 2,0 ml microcentrifuge tubes in numbered locations. Raised handle allows rack to be lifted without agitating samples.