ContactUsLinkComponent
Gas purifiers, AtmosPure™ Re-Gen
Gas purifiers, AtmosPure™ Re-Gen
Supplier:  LABCONCO
Certificates
undefined
Gas purifiers, AtmosPure™ Re-Gen
Supplier:  LABCONCO
Specifications
  • Catalog No:
  • LBCP5121831
  • LBCP5121835
  • Leak rating:
  • <1 x 10-6 ml/sec
  • <1 x 10-6 ml/sec
  • Inlet/outlet:
  • ¾" compression fittings
  • ¾" compression fittings
  • Weight:
  • 41 kg
  • 41 kg