Order Entry
Belgium
ContactUsLinkComponent
Refrigerator racks for centrifuge tubes, SP Bel-Art
Refrigerator racks for centrifuge tubes, SP Bel-Art
Catalog # BELAF189050050
undefined
Refrigerator racks for centrifuge tubes, SP Bel-Art
Catalog # BELAF189050050

Specifications

  • No. of Holes:
    28
  • Description:
    Refrigerator rack for 50 ml centrifuge tubes
  • W×D×H:
    320×200×150 mm
  • Pk:
    1

Specifications

About this item

PP

  • Temperature stabilised bench top storage
  • Flat perimeter rim provides space for labeling tape

Racks provides efficient cooling of centrifuge tubes in a cooling bath. The deep tray holds 2 liters of water and ice allows the tubes to rest on the bottom for maximum stability and exposure to the bath.