ContactUsLinkComponent
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Catalog # BELAH131990000
undefined
Autoclave deodoriser, triple strength, Odo-Clave®, SP Bel-Art
Catalog # BELAH131990000
Specifications
  • Description:
    Deodorant pads, 3× clove scent
  • Pk:
    100
Specifications
About this item
This heat-released deodorant pads neutralise autoclave odors with a pleasant scent.
  • Simply open foil pouch and place pad in bag prior to autoclaving
  • Pads have a three year life span; will last longer if stored in a sealed container
  • Triple strength clove releases three times the deodorant of the standard pads while costing less than two standard pads
W×L: 57×57 mm
Packaging: Packed 10 pads per bag, 10 bags per box.