Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Combustion calibration standard
Combustion calibration standard
Combustion calibration standard
Catalog # PERCN2410324
Supplier:
PerkinElmer
CAS Number:
Expand image
Combustion calibration standard
Catalog # PERCN2410324
Supplier:
PerkinElmer
CAS Number:
Specifications
Pk:
1
Description:
Combustion calibration statndard for sulphur analysis, cystine (26,69% S), solid 4 g
Specifications
About this item
Product Details
Product Details