ContactUsLinkComponent
Cannula with spray nozzle, Dosys™
Cannula with spray nozzle, Dosys™
Catalog # SOCO370.30500
Supplier:  Socorex Isba
undefined
Cannula with spray nozzle, Dosys™
Catalog # SOCO370.30500
Supplier:  Socorex Isba
Specifications
  • Ø×L:
    3×500 mm
  • Description:
    Cannula with spray nozzle, Luer-Lock
  • Material:
    Stainless steel
  • Pk:
    1
Specifications
About this item
This spray nozzle cannula fits on Luer-Lock self-refilling syringes.
Suitable where spraying and nebulising liquid is required.