Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Indium Standards
Single element standard for ICP, indium
Single element standard for ICP, indium
Single element standard for ICP, indium
Catalog # HEWL8500-6946
Supplier:
Agilent
CAS Number:
Expand image
Single element standard for ICP, indium
Catalog # HEWL8500-6946
Supplier:
Agilent
CAS Number:
Specifications
Concentration:
10 mg/L
Matrix:
2% HNO₃
Pk:
1
Element:
Indium
Symbol:
In
Specifications
About this item
Product Details
Product Details