ICP multi-element standards
Supplier: Agilent
Jump to:
About this item
4 Options Available Below
Product Details & Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- ConcentrationMatrixPkAvailabilityPrice
- HEWL8500-6940Multi-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, ZnMulti-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn10 mg/L5% HNO₃1Log in for availability1 * 1 ST704,00 €
- HEWL8500-6942Multi-element calibration standard-4, 100 ml, (12 elements in H20/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, ZrMulti-element calibration standard-4, 100 ml, (12 elements in H20/trace HF) B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr10 mg/L1% HNO₃ + (CHOH)₂(COOH)₂1Log in for availability1 * 1 ST385,00 €
- HEWL8500-6944Multi-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, YbMulti-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb10 mg/L5% HNO₃1Log in for availability1 * 1 ST291,00 €
- HEWL8500-6948Multi-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, SnMulti-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn10 mg/L10% HCl1Log in for availability1 * 1 ST561,00 €
This product has been blocked by your organization. Please contact your purchasing department for more information
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.