Orders LinkContactUsLinkComponent
Lambda DNA phage, <i>E. coli</i> W8850
Lambda DNA phage, E. coli W8850
DNA (lambda phage) E.coli W8850, 500 µG
Catalog # ROCKMB-104-0500
Lambda DNA phage, <i>E. coli</i> W8850
Catalog # ROCKMB-104-0500
CAS Number:  
Lambda DNA phage, E. coli W8850
DNA (lambda phage) E.coli W8850, 500 µG
CAS Number:  
Price
392,00 €
Log in for availability
1 * 500 µG
Product Details & Documents
Lambda phage consists of a virus particle including a head (also known as a capsid), a tail and tail fibres. The head contains the phage's double-stranded circular DNA genome. The phage particle recognises and binds to its host E.coli causing DNA in the head of the phage to be ejected through the tail into the cytoplasm of the bacterial cell. Lambda phage DNA is commonly used as a DNA molecular weight marker. Digestion of the lambda phage genome with HindIII produces a set of 8 fragments consisting of 125, 560, 2000, 2300, 4300, 9400, and 23000 base pairs.
Specifications
Specifications
  • Pk:
    500 µG
  • Description:
    DNA (lambda phage) <i>E.coli </i>W8850
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.