Order Entry
Belgium
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) (from calf thymus)
DNA (Deoxyribonucleic acid) (from calf thymus)
Catalog # ROCKMB-102-0100
CAS Number:  
DNA (Deoxyribonucleic acid) (from calf thymus)
Catalog # ROCKMB-102-0100
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
CAS Number:9007-49-2
 :100403-24-5
MDL Number:MFCD00130922