Order Entry
Belgium
Orders LinkContactUsLinkComponent
Density bottles, adjusted

Density bottles, adjusted

Cylindrical, class A, 24 ml, 24/15
Catalog # WITG8.126.139
Supplier:   witeg Labortechnik
Density bottles, adjusted
Catalog # WITG8.126.139
Supplier:  witeg Labortechnik
Density bottles, adjusted
Density bottles, adjusted

Cylindrical, class A, 24 ml, 24/15

Price
61,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Pycnometer, according to Hubbard

  • Adjusted to second decimal place

For use with bitumen and heavy oils.

Certifications: According to DIN 12806 and ASTM D-70

Specifications
  • NS:
    24/15
  • Volume (nom.):
    24 ml
  • Description:
    Cylindrical, class A
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.