Orders LinkContactUsLinkComponent
Draining racks, wall mounted
Draining racks, wall mounted
Draining rack, with 24 rods, 20 arches with 2 base pole connectors, 420×170×610 mm
Catalog # KOTT359-00028
Supplier:   KOETTERMANN
Draining racks, wall mounted
Catalog # KOTT359-00028
Supplier:  KOETTERMANN
Draining racks, wall mounted
Draining rack, with 24 rods, 20 arches with 2 base pole connectors, 420×170×610 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Wire, coated.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean
Specifications
  • Description:
    Draining rack, with 24 rods, 20 arches with 2 base pole connectors
  • W×D×H:
    420×170×610 mm