Orders LinkContactUsLinkComponent
Precision thermometer
Supplier:   Amarell
Precision thermometer
Supplier:  Amarell
Certificates
Precision thermometer
4 Options Available Below
Product Details & Documents
Dropping point thermometers according to Ubbelohde.
  • Richter-top finish
  • Colourless, prismatic capillary
  • Red or gallium filled
These enclosed milk glass scale thermometers are suitable for government verification or for government calibration.
 : Conforms to DIN 12785.
 : Available with or without verification certificate.
 : Packed in a transparent square plastic case.
Product Family Options
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARM235000-FL
    Dropping point thermometer
    Dropping point thermometer
    0…+110 °C
    ±1 °C
    240 mm
    Red special liquid
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,80 €
    63,80 €
    1 * 1 ST
       63,80 €
  • AMARM235000-FL/VC
    Dropping point thermometer, with verification certificate
    Dropping point thermometer, with verification certificate
    0…+110 °C
    ±1 °C
    240 mm
    Red special liquid
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €
  • AMARM235002-FL
    Dropping point thermometer
    Dropping point thermometer
    +50…+160 °C
    ±1 °C
    240 mm
    Red special liquid
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • AMARM235004-GA
    Dropping point thermometer
    Dropping point thermometer
    +100…+230 °C
    ±1 °C
    240 mm
    Gallium
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Metal case with metal ferrules, DIN 51801, mounted on dropping point therm
    Metal case with metal ferrules, DIN 51801, mounted on dropping point therm
    AMARM235030
    Log in for availability
    1 * 1 ST - 66,60 €