Order Entry
Belgium
ContactUsLinkComponent
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
  84871.320
 :  
undefined
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
  84871.320
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Nominal concentration (V/V):
    0.095 - 0.105 %
  • Evaporation residue:
    Max. 0.0001 %
  • Ca (Calcium):
    Max. 0.2 ppm
  • K (Potassium):
    Max. 0.2 ppm
  • Mg (Magnesium):
    Max. 0.2 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Transmittance (210 nm):
    Min. 20 %
  • Transmittance (220 nm):
    Min. 50 %
  • Transmittance (230 nm):
    Min. 80 %
  • Transmittance (240 nm):
    Min. 95 %
  • Transmittance (250 nm):
    Min. 99 %
  • Suitable for LC-MS:
    Passes test

 

 

 :
 :Packaged under nitrogen.
 :Ambient
 :200-929-3
 :76-05-1