Orders LinkContactUsLinkComponent
Shelving units
 :   Palbam
Shelving units
 :  Palbam
Shelving units
Shelving units
High-quality cleanroom shelving units on leveller legs with height adjustable feet or on white nylon 130 mm (5") cleanroom casters.
  • Shelf options: Solid or perforated
  • Shelves are removable or height adjustable every 200 mm (8")
  • 4 shelves per unit (extra shelves can be ordered separately)
Stainless steel 304, electropolished.
Product Family Options
Shelf units, solid
  • Description
    Material
    W×D×H
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1000×600×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1000×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on casters
    Stainless steel 304
    1000×400×1900 mm
    1 * 1 ST
    1 * 1 ST - 4 280,00 €
    4 280,00 €
    1 * 1 ST
       4 280,00 €
  • 4 solid shelves, on casters
    Stainless steel 304
    1000×600×1900 mm
    1 * 1 ST
    1 * 1 ST - 4 750,00 €
    4 750,00 €
    1 * 1 ST
       4 750,00 €
  • 4 solid shelves, on casters
    Stainless steel 304
    1200×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on casters
    Stainless steel 304
    1200×600×1900 mm
    1 * 1 ST
    1 * 1 ST - 5 140,00 €
    5 140,00 €
    1 * 1 ST
       5 140,00 €
    New Product
    New Product
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1200×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1400×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1500×600×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1500×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1600×600×1900 mm
    1 * 1 ST
    1 * 1 ST - 5 930,00 €
    5 930,00 €
    1 * 1 ST
       5 930,00 €
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1600×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 solid shelves, on leveller legs
    Stainless steel 304
    1600×400×1900 mm
    1 * 1 ST
    1 * 1 ST - 894,00 €
    894,00 €
    1 * 1 ST
       894,00 €
Shelf units, perforated
  • Description
    Material
    W×D×H
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1000×400×1900 mm
    1 * 1 ST
    1 * 1 ST - 4 410,00 €
    4 410,00 €
    1 * 1 ST
       4 410,00 €
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1000×600×1900 mm
    1 * 1 ST
    1 * 1 ST - 4 940,00 €
    4 940,00 €
    1 * 1 ST
       4 940,00 €
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1100×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on casters
    Stainless steel 304
    1200×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1200×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1300×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on casters
    Stainless steel 304
    1300×600×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on casters
    Stainless steel 304
    1400×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1500×400×1900 mm
    1 * 1 ST
    1 * 1 ST
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1600×400×1900 mm
    1 * 1 ST
    1 * 1 ST - 5 550,00 €
    5 550,00 €
    1 * 1 ST
       5 550,00 €
  • 4 perforated shelves, on leveller legs
    Stainless steel 304
    1600×600×1900 mm
    1 * 1 ST
    1 * 1 ST - 5 710,00 €
    5 710,00 €
    1 * 1 ST
       5 710,00 €