Orders LinkContactUsLinkComponent
Digital block heater, compact
Digital block heater, compact
Digital block heater, compact
Certificates
Digital block heater, compact
2 Options Available Below
Product Details & Documents
These versatile digital block heaters provide good temperature uniformity and stability ensuring reproducible results. Units are compact, light weight with digital temperature controls and display, they maximises valuable bench space. The units are ideal for sample preparation, enzyme preservation, DNA amplification and enzyme–substrate reactions.
  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combination of different size of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
 : Supplied with block extraction tool and plastic lid, but without blocks, these must be ordered separately.
 : Temp. stability value shown above is at 37 °C.
* RT = Ambient
** S - 1 block, 2×½ blocks, 4×¼ blocks or 1×½ block plus 2×¼ blocks
*** D - 2 blocks, 4×½ blocks, 8×¼ blocks or combination
Specifications for Product Family
Specifications
  • Catalog No:
  • 460-0284
  • 460-0283
  • Temperature range:
  • RT* +2...100 °C
  • RT* +2...100 °C
  • Temperature stability:
  • ≤±0,5 °C
  • ≤±0,5 °C
Specifications
Product Family Options
Product Information
  • Type
    No. of blocks
    Heating capacity
    Temperature Range
    Temp. stability
    W×D×H
    Weight
    Availability
    Price
  • Compact Dry Bath S**
    1
    58 W
    RT* +2...100 °C
    ≤±0,5 °C
    185×185×25 mm
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 400,00 €
    400,00 €
    1 * 1 ST
       400,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    460-0284
    Temperature range
    RT* +2...100 °C
    Temperature stability
    ≤±0,5 °C
  • Compact Dry Bath D***
    2
    77 W
    RT* +2...100 °C
    ≤±0,5 °C
    200×200×25 mm
    1,5 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 539,00 €
    539,00 €
    1 * 1 ST
       539,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    460-0283
    Temperature range
    RT* +2...100 °C
    Temperature stability
    ≤±0,5 °C
Accessories
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • ½ aluminium block
    ½ aluminium block
    460-0318
    50 ml tubes
    3
    Log in for availability
    1 * 1 ST - 85,80 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • ¼ aluminium block
    ¼ aluminium block
    460-0315
    1,5 ml tubes
    6
    Log in for availability
    1 * 1 ST - 24,70 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • ¼ aluminium block
    ¼ aluminium block
    460-0316
    2,0 ml tubes
    6
    Log in for availability
    1 * 1 ST - 24,70 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • ½ aluminium block
    ½ aluminium block
    460-0317
    15 ml tubes
    6
    Log in for availability
    1 * 1 ST - 85,80 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Plastic lid
    Plastic lid
    460-0323
    Compact D models
    -
    Log in for availability
    1 * 1 ST - 15,60 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Plastic lid
    Plastic lid
    460-0322
    Compact S models
    -
    Log in for availability
    1 * 1 ST - 9,50 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Heat insulator
    Heat insulator
    460-0321
    Compact D models
    -
    Log in for availability
    1 * 1 ST - 20,20 €
Product Information
  • Catalog #
    For
    W×D×H
    Availability
    Price
  • Full aluminium block
    Full aluminium block
    460-0319
    96-well microplate
    108×72×18 mm
    Log in for availability
    1 * 1 ST - 108,00 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • Heat insulator
    Heat insulator
    460-0320
    Compact S models
    -
    Log in for availability
    1 * 1 ST - 12,40 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • ¼ aluminium block
    ¼ aluminium block
    460-0314
    0,5 ml tubes
    12
    Log in for availability
    1 * 1 ST - 28,00 €
Product Information
  • Catalog #
    For
    No. of Holes
    Availability
    Price
  • ¼ aluminium block
    ¼ aluminium block
    460-0313
    0,2 ml Ø tubes
    24
    Log in for availability
    1 * 1 ST - 31,00 €