Orders LinkContactUsLinkComponent
Buckets
Buckets
Supplier:   hünersdorff GmbH
Buckets
Supplier:  hünersdorff GmbH
Certificates
Buckets
3 Options Available Below
Product Details & Documents
HDPE, translucent.
  • Spouted buckets for safe handling
  • Embossed scale and plastic handle
  • Food-safe
For industrial and laboratory applications.
Product Family Options
Product Information
  • Colour
    Capacity
    Ø
    Height
    Availability
    Price
  • Natural
    10,5 L
    286 mm
    283 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Black
    10,5 L
    286 mm
    283 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Natural
    17 L
    330 mm
    315 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST