Orders LinkContactUsLinkComponent
Capillary columns, GS-CarbonPLOT
Capillary columns, GS-CarbonPLOT
Supplier:   Agilent
Capillary columns, GS-CarbonPLOT
Supplier:  Agilent
Certificates
Capillary columns, GS-CarbonPLOT
Capillary columns, GS-CarbonPLOT
8 Options Available Below
Product Details & Documents
These columns are ideal for GC/MS.
  • High stability, carbon layer stationary phase
  • Unique selectivity for inorganic and organic gases
  • Extended temperature limit of 360 ºC
  • Retention stability not affected by water
Bonded monolithic carbon layer.
Similar phases: Carbopack, CLOT, Carboxen-1006 PLOT.
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • HEWL113-3112
    on 7" cage
    on 7" cage
    Capillary
    GS-CarbonPLOT
    1,50 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 552,00 €
    552,00 €
    1 * 1 ST
       552,00 €
  • HEWL115-3113
    on 7" cage
    on 7" cage
    Capillary
    GS-CarbonPLOT
    3,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 561,00 €
    561,00 €
    1 * 1 ST
       561,00 €
  • HEWL113-3132
    on 7" cage
    on 7" cage
    Capillary
    GS-CarbonPLOT
    1,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 941,00 €
    941,00 €
    1 * 1 ST
       941,00 €
  • HEWL113-3133
    on 7" cage
    on 7" cage
    Capillary
    GS-CarbonPLOT
    3,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 941,00 €
    941,00 €
    1 * 1 ST
       941,00 €
  • HEWL113-3133LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    GS-CarbonPLOT
    3,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL115-3133
    on 7" cage
    on 7" cage
    Capillary
    GS-CarbonPLOT
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 120,00 €
    1 120,00 €
    1 * 1 ST
       1 120,00 €
  • HEWL115-3133LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    GS-CarbonPLOT
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 400,00 €
    2 400,00 €
    1 * 1 ST
       2 400,00 €
  • HEWL113-3162
    on 7" cage
    on 7" cage
    Capillary
    GS-CarbonPLOT
    1,50 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 670,00 €
    1 670,00 €
    1 * 1 ST
       1 670,00 €