Orders LinkContactUsLinkComponent
Capillary columns, DB-624
Supplier:   Agilent
Capillary columns, DB-624
Supplier:  Agilent
Certificates
Capillary columns, DB-624
Capillary columns, DB-624
32 Options Available Below
Product Details & Documents
These columns are recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. They are specifically designed for the analysis of volatile priority pollutants and residual solvents.
  • Excellent inertness for active compounds
  • Bonded and cross-linked
  • Solvent rinsable
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 558-0375
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    1,12 µm
    0,20 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 703,00 €
    703,00 €
    1 * 1 ST
       703,00 €
  • HEWL123-1334E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    1,80 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €
  • 558-0368
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    1,80 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 931,00 €
    931,00 €
    1 * 1 ST
       931,00 €
  • 558-0372
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €
  • 558-0364
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    1,40 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • 558-0369
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    1,80 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 570,00 €
    1 570,00 €
    1 * 1 ST
       1 570,00 €
  • HEWL121-1324
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    1,00 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 672,00 €
    672,00 €
    1 * 1 ST
       672,00 €
  • HEWL122-1334
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    1,40 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 880,00 €
    880,00 €
    1 * 1 ST
       880,00 €
  • HEWL123-1364E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    1,80 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 770,00 €
    1 770,00 €
    1 * 1 ST
       1 770,00 €
  • HEWL124-1334
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    2,55 µm
    0,45 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €
  • HEWL124-1374
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    2,55 µm
    0,45 mm
    75 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 860,00 €
    1 860,00 €
    1 * 1 ST
       1 860,00 €
  • HEWL125-1334E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 210,00 €
    1 210,00 €
    1 * 1 ST
       1 210,00 €
  • HEWL125-1364
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 550,00 €
    1 550,00 €
    1 * 1 ST
       1 550,00 €
  • HEWL125-1374
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    75 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 920,00 €
    1 920,00 €
    1 * 1 ST
       1 920,00 €
  • HEWL122-1334E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    1,40 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 997,00 €
    997,00 €
    1 * 1 ST
       997,00 €
  • HEWL122-1364E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    1,40 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 600,00 €
    1 600,00 €
    1 * 1 ST
       1 600,00 €
  • HEWL125-1374E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    75 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 170,00 €
    2 170,00 €
    1 * 1 ST
       2 170,00 €
  • HEWL125-1364E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 760,00 €
    1 760,00 €
    1 * 1 ST
       1 760,00 €
  • HEWL128-1324E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    1,12 µm
    0,20 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 796,00 €
    796,00 €
    1 * 1 ST
       796,00 €
  • HEWL121-1324E
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    1,00 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 760,00 €
    760,00 €
    1 * 1 ST
       760,00 €
  • HEWL121-1324LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    1,00 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 910,00 €
    1 910,00 €
    1 * 1 ST
       1 910,00 €
  • HEWL122-1334LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    1,40 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 070,00 €
    2 070,00 €
    1 * 1 ST
       2 070,00 €
  • HEWL123-1334LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    1,80 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 180,00 €
    2 180,00 €
    1 * 1 ST
       2 180,00 €
  • HEWL124-1334LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    2,55 µm
    0,45 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWL125-13A4
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    100 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 590,00 €
    2 590,00 €
    1 * 1 ST
       2 590,00 €
  • HEWL125-13B4
    on 5" cage
    on 5" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    105 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 890,00 €
    2 890,00 €
    1 * 1 ST
       2 890,00 €
  • HEWL125-1314
    on 7" cage
    on 7" cage
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 726,00 €
    726,00 €
    1 * 1 ST
       726,00 €
  • HEWL125-1334LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 340,00 €
    2 340,00 €
    1 * 1 ST
       2 340,00 €
  • HEWL128-1314LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    1,12 µm
    0,20 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 620,00 €
    1 620,00 €
    1 * 1 ST
       1 620,00 €
  • HEWL128-1324LTM
    7890/6890 LTM module
    7890/6890 LTM module
    Capillary
    DB-624
    1,12 µm
    0,20 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 940,00 €
    1 940,00 €
    1 * 1 ST
       1 940,00 €
  • HEWL221-1324LTM
    5975T LTM toroid
    5975T LTM toroid
    Capillary
    DB-624
    1,00 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 150,00 €
    2 150,00 €
    1 * 1 ST
       2 150,00 €
  • HEWL222-1334LTM
    5975T LTM toroid
    5975T LTM toroid
    Capillary
    DB-624
    1,40 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,00 €
    2 240,00 €
    1 * 1 ST
       2 240,00 €