Capillary columns, DB-1
Supplier: Agilent
Certificates
About this item
These columns contain 100% dimethylpolysiloxane.
- Non-polar
- Very low bleed characteristics, ideal for GC/MS
Max. temperature: -60 to 325/350 °C
Frequently Bought Together
These columns contain 100% dimethylpolysiloxane.
Max. temperature: -60 to 325/350 °C