ContactUsLinkComponent
VWR® Traceable® logger trac, Temperature and Humidity Data Loggers
undefined
VWR® Traceable® logger trac, Temperature and Humidity Data Loggers
  • Catalog No:
  • 620-2325
  • 620-2326
  • Model:
  • Logger trac
  • Logger trac
  • Battery life:
  • 2 years
  • 2 years
  • Weight:
  • 70 g
  • 70 g
  • W×D×H:
  • 89×51×28 mm
  • 89×51×28 mm
  • Data memory - No. of readings:
  • 30000
  • 30000
Frequently Bought Together