Capillary columns, DB-35
Supplier: Agilent
Jump to:
About this item
They are solvent rinsable and very low bleed mid-polarity columns ideal for GC/MS. They are suitable for CLP pesticides.
- Excellent inertness for active compounds
- Enhanced mass spectral integrity
- Bonded and cross-linked
Methylpolysiloxane
Max. temperature: 40 to 320/340 °C
7 Options Available Below
Product Details & Documents
They are solvent rinsable and very low bleed mid-polarity columns ideal for GC/MS. They are suitable for CLP pesticides.
- Excellent inertness for active compounds
- Enhanced mass spectral integrity
- Bonded and cross-linked
Methylpolysiloxane
Max. temperature: 40 to 320/340 °C
Product Family Options
Product Information
- Column FormatPhaseFilm thicknessØ int.LengthAvailabilityPrice
- HEWL122-1932on 7" cage
- HEWL122-1962on 7" cage
- HEWL123-1932on 7" cage
- HEWL123-1933on 7" cage
- HEWL125-1912on 7" cage
- HEWL125-1932on 7" cage
- HEWL125-1937on 7" cage
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.