Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solution for chlorine determination

Buffer solution for chlorine determination

Supplier:   Hach
Buffer solution for chlorine determination
Supplier:  Hach
Certificates
Buffer solution for chlorine determination
Buffer solution for chlorine determination

2 Options Available Below

Product Details & Documents

For determination of chlorine by amperometric titration. APHA formulation 100 ml bottle.

Product Family Options

Product Information

  • pH value
    pH buffer type
    Pk
    Availability
    Price
  • 4,0
    Acetate
    100 ml
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 37,40 €
    37,40 €
    1 * 100 mL
       37,40 €

  • 7
    Phosphate
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,50 €
    32,50 €
    1 * 1 ST
       32,50 €

This product has been blocked by your organization. Please contact your purchasing department for more information
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.