Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH electrode filling solution

pH electrode filling solution

pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml, 65 mL
Catalog # HACH5194100
Supplier:   Hach
pH electrode filling solution
Catalog # HACH5194100
Supplier:  Hach
CAS Number:  
pH electrode filling solution

pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml, 65 mL

CAS Number:  
Price
21,80 €
Log in for availability
1 * 65 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    65 mL
  • Description:
    pH electrode filling solution, 4M KCl saturated with silver chloride (AgCl ), 65 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.