Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromophenol blue solution acid-base indicator

Bromophenol blue solution acid-base indicator

Dropping bottle, marked, 100 mL
Catalog # HACH1455232
Supplier:   Hach
Bromophenol blue solution acid-base indicator
Catalog # HACH1455232
Supplier:  Hach
CAS Number:  115-39-9
Bromophenol blue solution acid-base indicator
Bromophenol blue solution acid-base indicator

Dropping bottle, marked, 100 mL

CAS Number:  115-39-9
Price
28,40 €
Log in for availability
1 * 100 mL
Product Details & Documents

Bromphenol blue pH indicator solutions are used to determine acidity and alkalinity of water by titration method.

  • Visual transition interval: pH 3,0 (yellow) - pH 4,6 (blue)
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
Specifications

Specifications

  • Pack type:
    Dropping bottle, marked
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.