Orders LinkContactUsLinkComponent
pH buffer, single-use, SINGLET™
 :   Hach
pH buffer, single-use, SINGLET™
 :  Hach
pH buffer, single-use, SINGLET™
pH buffer, single-use, SINGLET™
These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.
  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0,02 at 25 °C
  • Traceable to NIST
Product Family Options
  • Description
    pH value
    pH accuracy
    Reference temperature
    Pack type
    Pk Info
    Pk
  • pH buffer solution, single-use
    4,01
    ±0,02
    25 °C
    Foil pouch
    20×20 ml pouches
    20
    1 * 20 ST
    1 * 20 ST - 50,70 €
    50,70 €
    1 * 20 ST
       50,70 €
  • pH buffer solution, single-use
    7,00
    ±0,02
    25 °C
    Foil pouch
    20×20 ml pouches
    20
    1 * 20 ST
    1 * 20 ST - 50,70 €
    50,70 €
    1 * 20 ST
       50,70 €
  • pH buffer solution, single-use
    10,01
    ±0,02
    25 °C
    Foil pouch
    20×20 ml pouches
    20
    1 * 20 ST
    1 * 20 ST - 50,70 €
    50,70 €
    1 * 20 ST
       50,70 €
Combination packs
  • Description
    pH value
    pH accuracy
    Reference temperature
    Pack type
    Pk Info
    Pk
  • pH buffer solution combination pack, single-use
    4,01/7,00
    Foil pouch
    20/pack (10 each of 4,01 pH and 7,00 pH)
    1 SET
    1 * 1 SET
    1 * 1 SET - 50,70 €
    50,70 €
    1 * 1 SET
       50,70 €
  • pH buffer solution combination pack, single-use
    7,00/10,01
    Foil pouch
    20/pack (10 each of 7,00 pH and 10,01 pH)
    1 SET
    1 * 1 SET
    1 * 1 SET - 50,70 €
    50,70 €
    1 * 1 SET
       50,70 €