ContactUsLinkComponent
Nitrites test kits, NI-15
Nitrites test kits, NI-15
Catalog # HACH2182000
Supplier:  Hach
CAS Number:  
undefined
Nitrites test kits, NI-15
Catalog # HACH2182000
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Nitrite test kit, LR, NI-15
  • Range:
    0 - 0,40 mg/l NO₂- N
Specifications
About this item
The accurate visual test kit method to determine nitrogen as nitrite by diazotisation of sample followed by colour comparison.
  • Simple and cost effective
  • Portable test kit
Delivery: Kit includes colour disc, comparator box and tubes, reagents ( but not deionised water), instructions and carrying case.