Order Entry
Belgium
ContactUsLinkComponent
Thymol blue solution in 2-propanol/water indicator
Thymol blue solution in 2-propanol/water indicator
Catalog # HACH25732
Supplier:  Hach
CAS Number:  
undefined
Thymol blue solution in 2-propanol/water indicator
Catalog # HACH25732
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
 :
 :466.6 g/mol
UN:1219
 :GHS02
 :GHS07
CAS Number:76-61-9
Formula:C₂₇H₃₀O₅S
MDL Number:MFCD00005869
Melting Pt:221
ADR:3 II
Merck Index:12,09541