ContactUsLinkComponent
Media set
Media set
Catalog # HACH2277700
Supplier:  Hach
CAS Number:  
undefined
Media set
Catalog # HACH2277700
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    3 Tests
  • Description:
    Total bacteria media set
  • For:
    Total Count
Specifications
About this item
Media set is a colorimetric method that is used for determination of total bacteria.
  • ToxTrak™ Toxicity test speeds sample evaluations
  • Economically alternative to bioassays
It includes dehydrated tryptic soy broth, screw cap vial and sterile transfer pippets.