ContactUsLinkComponent
Turbidity standard, ULR verification kit, Stablcal®
Turbidity standard, ULR verification kit, Stablcal®
Catalog # HACH2714600
Supplier:  Hach
CAS Number:  
undefined
Turbidity standard, ULR verification kit, Stablcal®
Catalog # HACH2714600
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Turbidity:
    1,0 NTU
    0,30 NTU
    0,10 NTU
    0,50 NTU
  • Description:
    ULR verification kit, Stablcal®
  • Pk Info:
    4x 100 ml
Specifications
About this item
Stablcal® verification kit containing 0.10 NTU, 0.30 NTU, 0.50 NTU, 1.0 NTU standards (100 ml each), sililcone oil, oiling cloth and instructions is used for verification purpose only.
  • Precise, pre-mixed formazin solutions
  • Enhanced stability saves time and labor
  • Confidence in results