ContactUsLinkComponent
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.1% indicator
Health
Corrosive
Environment
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.1% indicator
Supplier:  Hach
Certificates
undefined
Health
Corrosive
Environment
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.1% indicator
Supplier:  Hach
Certificates
Specification Test Results
  • Method:
    8149
  • Method Name:
    PAN
  • Parameter:
    Manganese
  • Platform :
    Solution
  • Range:
    0.006 - 0.700 mg/L Mn