Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Supplier:   Hach
Conductivity standards
Supplier:  Hach
Certificates
Conductivity standards
Conductivity standards
10 Options Available Below
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Product Family Options
Product Information
  • Conductivity
    Pk
    Availability
    Price
  • HACH2971849
    Sodium chloride standard solution, 49 mg/l NaCl
    Sodium chloride standard solution, 49 mg/l NaCl
    100 µS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 52,00 €
    52,00 €
    1 * 500 mL
       52,00 €
  • HACHLZW9700.99
    Conductivity standard solution
    Conductivity standard solution
    147 µS/cm
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €
  • HACH2307542
    Conductivity standard solution, 85,47 mg/l TDS
    Conductivity standard solution, 85,47 mg/l TDS
    180 µS/cm
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 32,90 €
    32,90 €
    1 * 100 mL
       32,90 €
  • HACH1440026
    Sodium chloride standard solution, pour-top, 491 mg/l NaCl
    Sodium chloride standard solution, pour-top, 491 mg/l NaCl
    1000 µS/cm
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 16,20 €
    16,20 €
    1 * 50 mL
       16,20 €
  • HACH1440042
    Sodium chloride standard solution, 491 mg/l Nacl
    Sodium chloride standard solution, 491 mg/l Nacl
    1000 µS/cm
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 31,30 €
    31,30 €
    1 * 100 mL
       31,30 €
  • HACH1440049
    Sodium chloride standard solution, 491 mg/l NaCl
    Sodium chloride standard solution, 491 mg/l NaCl
    1000 µS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 74,70 €
    74,70 €
    1 * 500 mL
       74,70 €
  • HACHLZW9710.99
    Conductivity standard solution
    Conductivity standard solution
    1413 µS/cm
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €
  • HACH210542
    Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl
    Sodium chloride standard solution, 995 mg/l TDS, 1000 mg/l Nacl
    1990 µS/cm
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 33,60 €
    33,60 €
    1 * 100 mL
       33,60 €
  • HACHLZW9720.99
    Conductivity standard solution
    Conductivity standard solution
    12,88 mS/cm
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 25,20 €
    25,20 €
    1 * 250 mL
       25,20 €
  • HACH2307442
    Sodium chloride standard solution, 10 246 mg/l NaCl
    Sodium chloride standard solution, 10 246 mg/l NaCl
    18 000 µS/cm
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 31,30 €
    31,30 €
    1 * 100 mL
       31,30 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.