Order Entry
Belgium
Orders LinkContactUsLinkComponent
Chloride standard solution

Chloride standard solution

Chloride standard solution, 1000 mg/l, 500 mL
Catalog # HACH18349
Supplier:   Hach
Chloride standard solution
Catalog # HACH18349
Supplier:  Hach
CAS Number:  
Chloride standard solution
Chloride standard solution

Chloride standard solution, 1000 mg/l, 500 mL

CAS Number:  
Price
74,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

For accuracy checks in chloride determinations. Also suitable for standardising conductivity apparatus.

Documents

Specifications

Specifications

  • Concentration:
    1000 mg/l
  • Pk:
    500 mL
  • Description:
    Chloride standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.