Order Entry
Belgium
Orders LinkContactUsLinkComponent
Fluoride standard solution

Fluoride standard solution

Fluoride standard solution, 5 mg/l as F, 500 mL
Catalog # HACH2797149
Supplier:   Hach
Fluoride standard solution
Catalog # HACH2797149
Supplier:  Hach
CAS Number:  
Fluoride standard solution
Fluoride standard solution

Fluoride standard solution, 5 mg/l as F, 500 mL

CAS Number:  
Price
51,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

For accuracy checks in fluoride determinations.

Specifications

Specifications

  • Concentration:
    5 mg/l as F
  • Pk:
    500 mL
  • Description:
    Fluoride standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.