Orders LinkContactUsLinkComponent
Phosphate standard solutions
Phosphate standard solutions
Supplier:   Hach
Phosphate standard solutions
Supplier:  Hach
Certificates
Phosphate standard solutions
Phosphate standard solutions
8 Options Available Below
Product Details & Documents
Standard solution for accuracy checks in phosphate (phosphorus) determinations.
Product Family Options
Product Information
  • Concentration
    Pk
    Availability
    Price
  • HACH256949
    Phosphate standard solution, 500 ml (NIST)
    Phosphate standard solution, 500 ml (NIST)
    1 mg/l as PO43-
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 40,60 €
    40,60 €
    1 * 500 mL
       40,60 €
  • HACH2059716
    Phosphate standard solution, 946 ml (NIST)
    Phosphate standard solution, 946 ml (NIST)
    3 mg/l as PO43-
    946 mL
    Log in for availability
    Log in for availability
    1 * 946 mL
    1 * 946 mL - 69,80 €
    69,80 €
    1 * 946 mL
       69,80 €
  • HACH1420416
    Phosphate standard solution, 946 ml (NIST)
    Phosphate standard solution, 946 ml (NIST)
    10 mg/l as PO4
    946 mL
    Log in for availability
    Log in for availability
    1 * 946 mL
    1 * 946 mL - 59,80 €
    59,80 €
    1 * 946 mL
       59,80 €
  • HACH1424342
    Phosphate standard solution, 100 ml (NIST)
    Phosphate standard solution, 100 ml (NIST)
    15 mg/l as PO4
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 50,10 €
    50,10 €
    1 * 100 mL
       50,10 €
  • HACH17120H
    Phosphate standard solution, pack of 20×2 ml ampoules (NIST)
    Phosphate standard solution, pack of 20×2 ml ampoules (NIST)
    50 mg/l as PO4
    20
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 70,80 €
    70,80 €
    1 * 20 ST
       70,80 €
  • 705-0076
    Phosphate standard solution, pack of 16×10 ml voluette ampoules (NIST)
    Phosphate standard solution, pack of 16×10 ml voluette ampoules (NIST)
    50 mg/l as PO4
    16
    Log in for availability
    Log in for availability
    1 * 16 ST
    1 * 16 ST - 70,10 €
    70,10 €
    1 * 16 ST
       70,10 €
  • HACH17149
    Phosphate standard solution, 500 ml (NIST)
    Phosphate standard solution, 500 ml (NIST)
    50 mg/l as PO4
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 49,10 €
    49,10 €
    1 * 500 mL
       49,10 €
  • HACH1436832
    Phosphate standard solution, 100 ml (NIST)
    Phosphate standard solution, 100 ml (NIST)
    100 mg/l as PO4
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 28,40 €
    28,40 €
    1 * 100 mL
       28,40 €