Orders LinkContactUsLinkComponent
VWR®, Cell Strainers
VWR®, Cell Strainers
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, Cell Strainers
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, Cell Strainers
3 Options Available Below
Product Details & Documents
Manufactured from a strong nylon mesh with evenly spaced mesh pores, these cell strainers are sterile, easy to use devices for quickly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Designed to protect valuable flow cytometry and cell sorting instrumentation by reliably removing clumps and debris from cell suspensions and clinical samples prior to analysis.
  • Available in three mesh sizes with different colours for easy identification
  • Extended lip on the strainer enables aseptic handling with forceps
  • Ready to use, sterilised by electron beam irradiation
  • Individually wrapped
  • Fit into most 50 ml conical tubes
  • Free from DNase and RNase, non pyrogenic
ISO 9001, ISO 13485
Certifications: ISO 9001, ISO 13485
Specifications for Product Family
Specifications
  • Catalog No:
  • 732-2757
  • 732-2758
  • 732-2759
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 732-2757
    Cell strainers, 40 µm pore size, blue frame
    Cell strainers, 40 µm pore size, blue frame
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 113,00 €
    113,00 €
    1 * 50 ST
       113,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-2757
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 732-2758
    Cell strainers, 70 µm pore size, white frame
    Cell strainers, 70 µm pore size, white frame
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 113,00 €
    113,00 €
    1 * 50 ST
       113,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-2758
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 732-2759
    Cell strainers, 100 µm pore size, yellow frame
    Cell strainers, 100 µm pore size, yellow frame
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 113,00 €
    113,00 €
    1 * 50 ST
       113,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-2759
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact