Orders LinkContactUsLinkComponent
Eppendorf® Assay/Reader Microplates
   Sustainable Options Available
Supplier:   EPPENDORF
Eppendorf® Assay/Reader Microplates
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Eppendorf® Assay/Reader Microplates
8 Options Available Below
Product Details & Documents
Eppendorf Assay/Reader plates are designed for diverse microplate assays, optimising absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximising reflection
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes
  • All plates are optimised for minimal autofluorescence and autoluminescence
  • Available in lot-certified PCR clean purity grade
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability
Perfect plate for your microplate assay protocol. Eppendorf assay plate range has the plate for your application, optimised for measuring absorbance in the visible range, fluorescence and chemiluminescence. The range includes both solid plates for top-reading plate readers and versatile clear and black/clear plates for use with both top- and bottom-reading plate readers.
Experience clear signals even with low-concentration samples. Black microplates are ideal for fluorescence detection, with an excellent signal-to-noise ratio, providing low detection limits in assays. White microplates maximise reflection for high-sensitivity detection of luminescence or weak fluorescence.
Specifications for Product Family
Specifications
  • Catalog No:
  • 737-0169
  • 737-0170
  • 737-0171
  • 737-0172
  • 737-0248
  • 737-0249
  • 737-0227
  • 737-0228
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
96-well
Product Information
  • Colour
    No. of wells
    Sterile
    Availability
    Price
  • 737-0169
    U-bottom, PCR clean
    U-bottom, PCR clean
    White/grey
    96
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 354,00 €
    354,00 €
    1 * 80 ST
       354,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0169
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0170
    V-bottom, PCR clean
    V-bottom, PCR clean
    White/grey
    96
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 354,00 €
    354,00 €
    1 * 80 ST
       354,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0170
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0171
    U-bottom, PCR clean
    U-bottom, PCR clean
    Black/white
    96
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 354,00 €
    354,00 €
    1 * 80 ST
       354,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0171
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0172
    V-bottom, PCR clean
    V-bottom, PCR clean
    Black/white
    96
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 354,00 €
    354,00 €
    1 * 80 ST
       354,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0172
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0248
    Flat bottom, PCR clean
    Flat bottom, PCR clean
    White/grey
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 354,00 €
    354,00 €
    Log in for availability
    Each
    Each - 347,00 €
    347,00 €
    1 * 80 ST
       354,00 €
    Each
       347,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0248
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0249
    Flat bottom, PCR clean
    Flat bottom, PCR clean
    Black/white
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 354,00 €
    354,00 €
    1 * 80 ST
       354,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0249
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
384-well
Product Information
  • Colour
    No. of wells
    Sterile
    Availability
    Price
  • 737-0227
    V-bottom, PCR clean
    V-bottom, PCR clean
    White/grey
    384
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 908,00 €
    908,00 €
    1 * 80 ST
       908,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0227
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0228
    V-bottom, PCR clean
    V-bottom, PCR clean
    Black/white
    384
    -
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 908,00 €
    908,00 €
    1 * 80 ST
       908,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0228
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact