Nitrogen-nitrate standard solution
Supplier: Hach
Jump to:
About this item
Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.
4 Options Available Below
Product Details & Documents
Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- ConcentrationPkAvailabilityPrice
- HACH204649Nitrogen-nitrate standard solutionNitrogen-nitrate standard solution1 mg/l as NO₃-N (NIST)500 mLLog in for availability1 * 500 mL44,90 €
- HACH2415132Nitrogen-nitrate standard solutionNitrogen-nitrate standard solution15 mg/l as NO₃-N (NIST)100 mLLog in for availability1 * 100 mL20,40 €
- 705-2025Nitrogen-nitrate standard solutionNitrogen-nitrate standard solution100 mg/l as NO₃-N (NIST)500 mLLog in for availability1 * 500 mL53,60 €
- 705-2010Nitrogen-nitrate standard solution, pack of 16×10 ml voluette ampoulesNitrogen-nitrate standard solution, pack of 16×10 ml voluette ampoules500 mg/l as NO₃-N (NIST)16Log in for availability1 * 16 ST70,10 €