ContactUsLinkComponent
Visual colour kits, swimming pools, Pool Master
Catalog # HACH2315001
Supplier:  Hach
CAS Number:  
undefined
Visual colour kits, swimming pools, Pool Master
Catalog # HACH2315001
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Pool master test kit
Specifications
About this item
Pool Master test kit combines several tests into a single test kit. The test kit includes tests for alkalinity (20 to 400 mg/L), free and total chlorine (0 to 3,4 mg/L), hardness (20 to 400 mg/L), and pH (6,6 to 8,4 mg/L) and specialised tests for total alkalinity, calcium hardness, and cyanuric acid.
Delivery: Kit includes ten reagents, clippers, graduated cylinder, sample cell with cap, dipstick, two colour discs, viewing tubes, comparator box, instructions and carrying case.