Order Entry
Belgium
ContactUsLinkComponent
Visual comparator test kits, nitrate, NI-11
Visual comparator test kits, nitrate, NI-11
Catalog # HACH146803
Supplier:  Hach
CAS Number:  
undefined
Visual comparator test kits, nitrate, NI-11
Catalog # HACH146803
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Colour disc, model NI-11
  • Type:
    Cadmium reduction
  • Range:
    0 - 40 mg/l

Specifications

About this item

Simple, low cost, portable, colour disc method of testing nitrate.

Delivery: Kit includes colour disc, comparator box and tubes, reagents, instructions and carrying case.