ContactUsLinkComponent
Visual colour comparator kit, phosphate
Visual colour comparator kit, phosphate
Catalog # HACH224902
Supplier:  Hach
CAS Number:  
undefined
Visual colour comparator kit, phosphate
Catalog # HACH224902
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Phosphate test kit, (ortho+meta), model PO-23
  • Range:
    0,1 - 4 mg/l
    1 - 40 mg/l
Specifications
About this item
Simple, economical rapid colour disc method with an easy to carry test kit.
Delivery: Kit includes three reagents, colour disc, comparator box, two viewing tubes, bottle, dropper tube, stove and cover, test tube clamp, deionised water, instructions and carrying case.